siRNA Details
| Gene Symbol: Esp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244763.1 | NP_001231692.1 | 2 | 174 | ||
| GenBank mRNA | AB306997.1 | - | 2 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esp18| Gene Symbol: Esp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244763.1 | NP_001231692.1 | 2 | 174 | ||
| GenBank mRNA | AB306997.1 | - | 2 | 174 | ||