siRNA Details
| Gene Symbol: Esp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177584.1 | NP_001171055.1 | 3 | 703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esp8| Gene Symbol: Esp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177584.1 | NP_001171055.1 | 3 | 703 | ||