siRNA Details
| Gene Symbol: Esp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177582.1 | NP_001171053.1 | 1 | 135 | ||
| GenBank mRNA | AB307002.1 | - | 1 | 135 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esp23| Gene Symbol: Esp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177582.1 | NP_001171053.1 | 1 | 135 | ||
| GenBank mRNA | AB307002.1 | - | 1 | 135 | ||