siRNA Details
| Gene Symbol: ND4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140187.1 | - | 1 | 153 | ||
| AK144853.1 | - | 153 | ||||
| AK153747.1 | - | 152 | ||||
| AK157367.1 | - | 153 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ND4L| Gene Symbol: ND4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140187.1 | - | 1 | 153 | ||
| AK144853.1 | - | 153 | ||||
| AK153747.1 | - | 152 | ||||
| AK157367.1 | - | 153 | ||||