siRNA Details
| Gene Symbol: AI481877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135748.1 | - | 5 | 449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI481877| Gene Symbol: AI481877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135748.1 | - | 5 | 449 | ||