siRNA Details
| Gene Symbol: Gm17359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142953.1 | NP_001136425.1 | 4 | 490 | ||
| GenBank mRNA | BU583367.1 | - | 3 | 257 | ||
| CN840131.1 | - | 493 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17359| Gene Symbol: Gm17359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142953.1 | NP_001136425.1 | 4 | 490 | ||
| GenBank mRNA | BU583367.1 | - | 3 | 257 | ||
| CN840131.1 | - | 493 | ||||