siRNA Details
| Gene Symbol: Gm10778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142963.1 | NP_001136435.1 | 4 | 2856 | ||
| GenBank mRNA | BC057364.1 | - | 4 | 2856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10778| Gene Symbol: Gm10778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142963.1 | NP_001136435.1 | 4 | 2856 | ||
| GenBank mRNA | BC057364.1 | - | 4 | 2856 | ||