siRNA Details
| Gene Symbol: Gm17660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163772.1 | NP_001157244.1 | 4 | 181 | ||
| GenBank mRNA | EU642618.1 | - | 4 | 181 | ||
| HM015622.1 | - | 4 | 209 | |||
| HM015623.1 | - | 4 | 165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17660| Gene Symbol: Gm17660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163772.1 | NP_001157244.1 | 4 | 181 | ||
| GenBank mRNA | EU642618.1 | - | 4 | 181 | ||
| HM015622.1 | - | 4 | 209 | |||
| HM015623.1 | - | 4 | 165 | |||