siRNA Details
| Gene Symbol: Gm15881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177534.2 | NP_001171005.1 | 1 | 282 | ||
| GenBank mRNA | EU477373.1 | - | 1 | 207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15881| Gene Symbol: Gm15881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177534.2 | NP_001171005.1 | 1 | 282 | ||
| GenBank mRNA | EU477373.1 | - | 1 | 207 | ||