siRNA Details
| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159743.1 | NP_001153215.1 | 2 | 755 | ||
| GenBank mRNA | AK053903.1 | - | 2 | 639 | ||
| AK134499.1 | - | 2 | 758 | |||
| BC046522.1 | - | 2 | 738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam150b| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159743.1 | NP_001153215.1 | 2 | 755 | ||
| GenBank mRNA | AK053903.1 | - | 2 | 639 | ||
| AK134499.1 | - | 2 | 758 | |||
| BC046522.1 | - | 2 | 738 | |||