siRNA Details
Gene Symbol: Gm17455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164374.1 | NP_001157846.1 | 1 | 239 | ||
GenBank mRNA | BE650548.1 | - | 1 | 239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm17455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164374.1 | NP_001157846.1 | 1 | 239 | ||
GenBank mRNA | BE650548.1 | - | 1 | 239 |