siRNA Details
| Gene Symbol: Vmn1r221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167542.1 | NP_001161014.1 | 1 | 885 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r221| Gene Symbol: Vmn1r221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167542.1 | NP_001161014.1 | 1 | 885 | ||