siRNA Details
Gene Symbol: Gm17689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167584.1 | NP_001161056.1 | 3 | 293 | ||
GenBank mRNA | EU703627.1 | - | 3 | 293 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm17689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167584.1 | NP_001161056.1 | 3 | 293 | ||
GenBank mRNA | EU703627.1 | - | 3 | 293 |