siRNA Details
| Gene Symbol: Gm17365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167592.2 | NP_001161064.1 | 3 | 956 | ||
| GenBank mRNA | EU004071.1 | - | 3 | 956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17365| Gene Symbol: Gm17365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167592.2 | NP_001161064.1 | 3 | 956 | ||
| GenBank mRNA | EU004071.1 | - | 3 | 956 | ||