siRNA Details
| Gene Symbol: Gm17727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167673.1 | NP_001161145.1 | 2 | 325 | ||
| Gene Symbol: D730048I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136605.1 | - | 586 | |||

Gm17727
Genomic Map