siRNA Details
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017182.2 | NP_058878.1 | 7 | 907 | ||
| XM_006253573.2 | XP_006253635.1 | 7 | 945 | |||
| XM_006253574.2 | XP_006253636.1 | 7 | 1035 | |||
| XM_006253575.2 | XP_006253637.1 | 7 | 936 | |||
| XM_006253576.2 | XP_006253638.1 | 7 | 933 | |||
| XM_006253577.2 | XP_006253639.1 | 7 | 945 | |||
| GenBank mRNA | BC089093.1 | - | 7 | 907 | 7126329 | |
| M99065.2 | - | 6 | 748 | |||
| U79139.1 | - | 7 | 852 | |||

H2afy
Genomic Map