siRNA Details
Gene Symbol: 4930469G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195189.1 | NP_001182118.1 | 1 | 375 | ||
GenBank mRNA | AK015528.1 | - | 375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930469G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195189.1 | NP_001182118.1 | 1 | 375 | ||
GenBank mRNA | AK015528.1 | - | 375 |