siRNA Details
| Gene Symbol: Gm19402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205032.1 | NP_001191961.1 | 1 | 30 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19402| Gene Symbol: Gm19402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205032.1 | NP_001191961.1 | 1 | 30 | ||