siRNA Details
| Gene Symbol: Epg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195633.1 | NP_001182562.1 | 44 | 9525 | ||
| GenBank mRNA | AK017297.1 | - | 1 | 1138 | ||
| AK122536.1 | - | 29 | 6509 | |||
| BC033351.1 | - | 8 | 3226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epg5| Gene Symbol: Epg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195633.1 | NP_001182562.1 | 44 | 9525 | ||
| GenBank mRNA | AK017297.1 | - | 1 | 1138 | ||
| AK122536.1 | - | 29 | 6509 | |||
| BC033351.1 | - | 8 | 3226 | |||