siRNA Details
| Gene Symbol: Epg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195633.1 | NP_001182562.1 | 23, 24 | 4226 | ||
| GenBank mRNA | AK028969.1 | - | 19, 20 | 2752 | ||
| AK122536.1 | - | 8, 9 | 1210 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epg5| Gene Symbol: Epg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195633.1 | NP_001182562.1 | 23, 24 | 4226 | ||
| GenBank mRNA | AK028969.1 | - | 19, 20 | 2752 | ||
| AK122536.1 | - | 8, 9 | 1210 | |||