siRNA Details
| Gene Symbol: Gm38425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251890.1 | NP_001238819.1 | 3 | 870 | ||
| GenBank mRNA | DV646607.1 | - | 1 | 323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm38425| Gene Symbol: Gm38425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251890.1 | NP_001238819.1 | 3 | 870 | ||
| GenBank mRNA | DV646607.1 | - | 1 | 323 | ||