siRNA Details
| Gene Symbol: Gm4980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195529.1 | NP_001182458.1 | 1 | 1986 | ||
| GenBank mRNA | AK032609.1 | - | 1 | 1059 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4980| Gene Symbol: Gm4980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195529.1 | NP_001182458.1 | 1 | 1986 | ||
| GenBank mRNA | AK032609.1 | - | 1 | 1059 | ||