siRNA Details
Gene Symbol: Adra1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017191.2 | NP_058887.2 | 2 | 1249 | ||
XM_006252107.2 | XP_006252169.1 | 3 | 2234 | |||
XM_006252108.2 | XP_006252170.1 | 4 | 2139 | |||
XM_006252109.2 | XP_006252171.1 | 3 | 2233 | |||
XM_008770744.1 | XP_008768966.1 | 3 | 2234 | |||
GenBank mRNA | U07126.1 | - | 2 | 1249 | ||
U13368.1 | - | 2 | 1269 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
