siRNA Details
| Gene Symbol: Sp3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005870.1 | - | 3 | 284 | ||
| AK013593.1 | - | 3 | 243 | |||
| BY152393.1 | - | 3 | 294 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183265.1 | - | 284 | Chr. - - [-] on Build | ||

Sp3os
Genomic Map