siRNA Details
| Gene Symbol: Gm15056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177471.1 | NP_001170942.1 | 2, 3 | 149 | ||
| GenBank mRNA | DQ012052.1 | - | 1, 2 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15056| Gene Symbol: Gm15056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177471.1 | NP_001170942.1 | 2, 3 | 149 | ||
| GenBank mRNA | DQ012052.1 | - | 1, 2 | 89 | ||