siRNA Details
| Gene Symbol: Gm6537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195091.1 | NP_001182020.1 | 3, 4 | 206 | ||
| GenBank mRNA | AI427275.1 | - | 3, 4 | 206 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6537| Gene Symbol: Gm6537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195091.1 | NP_001182020.1 | 3, 4 | 206 | ||
| GenBank mRNA | AI427275.1 | - | 3, 4 | 206 | ||