siRNA Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181990.2 | NP_871790.2 | 4 | 970 | ||
| GenBank mRNA | AY369078.2 | - | 517 | |||
| BC145587.1 | - | 4 | 970 | 9054153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmtm1| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181990.2 | NP_871790.2 | 4 | 970 | ||
| GenBank mRNA | AY369078.2 | - | 517 | |||
| BC145587.1 | - | 4 | 970 | 9054153 | ||