siRNA Details
| Gene Symbol: Arhgap33os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC065044.1 | - | 12 | 2082 | 5685982 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap33os| Gene Symbol: Arhgap33os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC065044.1 | - | 12 | 2082 | 5685982 | |