siRNA Details
| Gene Symbol: Tfpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177321.1 | NP_001170792.1 | 7 | 717 | ||
| NM_017200.1 | NP_058896.1 | 6 | 645 | |||
| XM_006234440.2 | XP_006234502.1 | 7 | 940 | |||
| XM_006234441.2 | XP_006234503.1 | 7 | 1961 | |||
| XM_006234444.2 | XP_006234506.1 | 7 | 1974 | |||
| XM_008761980.1 | XP_008760202.1 | 7 | 959 | |||
| XM_008761981.1 | XP_008760203.1 | 7 | 723 | |||
| XM_008761982.1 | XP_008760204.1 | 7 | 953 | |||
| XM_008761983.1 | XP_008760205.1 | 7 | 1976 | |||
| XM_008761984.1 | XP_008760206.1 | 7 | 1989 | |||
| XM_008761985.1 | XP_008760207.1 | 7 | 965 | |||
| XM_008761986.1 | XP_008760208.1 | 7 | 959 | |||
| GenBank mRNA | BC087020.1 | - | 7 | 717 | 7192382 | |
| D10926.1 | - | 6 | 645 | |||
| FQ219876.1 | - | 4 | 351 | |||
| FQ220339.1 | - | 4 | 345 | |||
| FQ220655.1 | - | 4 | 442 | |||
| FQ220711.1 | - | 4 | 442 | |||
| FQ229364.1 | - | 4 | 374 | |||

Tfpi
Genomic Map