siRNA Details
| Gene Symbol: Spin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243002.1 | NP_001229931.1 | 2 | 906 | ||
| GenBank mRNA | AK015110.1 | - | 2 | 908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spin2d| Gene Symbol: Spin2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243002.1 | NP_001229931.1 | 2 | 906 | ||
| GenBank mRNA | AK015110.1 | - | 2 | 908 | ||