siRNA Details
Gene Symbol: Spin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243002.1 | NP_001229931.1 | 2 | 992 | ||
GenBank mRNA | AK015110.1 | - | 2 | 994 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243002.1 | NP_001229931.1 | 2 | 992 | ||
GenBank mRNA | AK015110.1 | - | 2 | 994 |