siRNA Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033202.3 | NP_001028374.1 | 13 | 2266 | ||
| GenBank mRNA | AK078164.1 | - | 9 | 1618 | ||
| AK144578.1 | - | 13 | 2267 | |||
| AK162153.1 | - | 1315 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp30| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033202.3 | NP_001028374.1 | 13 | 2266 | ||
| GenBank mRNA | AK078164.1 | - | 9 | 1618 | ||
| AK144578.1 | - | 13 | 2267 | |||
| AK162153.1 | - | 1315 | ||||