siRNA Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017206.1 | NP_058902.1 | 6, 7 | 866 | ||
| XM_006236903.2 | XP_006236965.1 | 5, 6 | 975 | |||
| XM_008763123.1 | XP_008761345.1 | 7, 8 | 1136 | |||
| GenBank mRNA | M96601.1 | - | 6, 7 | 866 | ||

Slc6a6
Genomic Map