siRNA Details
| Gene Symbol: Zfp575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033205.3 | NP_001028377.1 | 4 | 2316 | ||
| GenBank mRNA | AK159025.1 | - | 4 | 2317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp575| Gene Symbol: Zfp575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033205.3 | NP_001028377.1 | 4 | 2316 | ||
| GenBank mRNA | AK159025.1 | - | 4 | 2317 | ||