siRNA Details
| Gene Symbol: Zfp575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033205.3 | NP_001028377.1 | 4 | 1301 | ||
| GenBank mRNA | AK138180.1 | - | 4 | 1260 | ||
| AK159025.1 | - | 4 | 1302 | |||
| BC150867.1 | - | 4 | 1267 | 9087778 | ||
| BC150873.1 | - | 4 | 1267 | 9087784 | ||

Zfp575
Genomic Map