siRNA Details
| Gene Symbol: Dcun1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024504.2 | NP_001019675.2 | 7 | 2692 | ||
| NM_001042649.1 | NP_001036114.1 | 6 | 2609 | |||
| NM_001042650.1 | NP_001036115.1 | 5 | 2512 | |||
| NM_001042651.1 | NP_001036116.1 | 6 | 2595 | |||
| GenBank mRNA | AK040130.1 | - | 3 | 2124 | ||
| AK154550.1 | - | 6 | 2615 | |||
| AK155726.1 | - | 7 | 2408 | |||
| AK161694.1 | - | 6 | 2301 | |||
| BC030839.1 | - | 7 | 2400 | |||
| BC052676.1 | - | 5 | 2514 | |||

Dcun1d2
Genomic Map