siRNA Details
| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034895.3 | NP_001030067.2 | 13 | 1973 | ||
| GenBank mRNA | AK033649.1 | - | 6 | 722 | ||
| AK122438.1 | - | 6 | 3581 | |||
| BC072571.1 | - | 12 | 2642 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfr2| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034895.3 | NP_001030067.2 | 13 | 1973 | ||
| GenBank mRNA | AK033649.1 | - | 6 | 722 | ||
| AK122438.1 | - | 6 | 3581 | |||
| BC072571.1 | - | 12 | 2642 | |||