siRNA Details
| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045515.1 | NP_001038980.1 | 33 | 6808 | ||
| NM_001164483.1 | NP_001157955.1 | 32 | 6877 | |||
| GenBank mRNA | AB093272.1 | - | 32 | 6694 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Synj1| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045515.1 | NP_001038980.1 | 33 | 6808 | ||
| NM_001164483.1 | NP_001157955.1 | 32 | 6877 | |||
| GenBank mRNA | AB093272.1 | - | 32 | 6694 | ||