siRNA Details
| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045515.1 | NP_001038980.1 | 4 | 497 | ||
| NM_001164483.1 | NP_001157955.1 | 4 | 590 | |||
| GenBank mRNA | AB093272.1 | - | 3 | 383 | ||
| AK080946.1 | - | 4 | 464 | |||

Synj1
Genomic Map