siRNA Details
| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081060.1 | NP_001074529.1 | 11 | 1681 | ||
| GenBank mRNA | AF139194.1 | - | 1 | 32 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a3| Gene Symbol: Slc9a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081060.1 | NP_001074529.1 | 11 | 1681 | ||
| GenBank mRNA | AF139194.1 | - | 1 | 32 | ||