siRNA Details
| Gene Symbol: E330034G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033214.2 | NP_001028386.1 | 7 | 1444 | ||
| GenBank mRNA | AK087874.1 | - | 7 | 1245 | ||
| AK136036.1 | - | 7 | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E330034G19Rik| Gene Symbol: E330034G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033214.2 | NP_001028386.1 | 7 | 1444 | ||
| GenBank mRNA | AK087874.1 | - | 7 | 1245 | ||
| AK136036.1 | - | 7 | 1445 | |||