siRNA Details
| Gene Symbol: E330034G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033214.2 | NP_001028386.1 | 1 | 88 | ||
| GenBank mRNA | AK136036.1 | - | 1 | 89 | ||
| BC139082.1 | - | 1 | 59 | 8862104 | ||
| BC139083.1 | - | 1 | 59 | 8862105 | ||

E330034G19Rik
Genomic Map