siRNA Details
| Gene Symbol: E330034G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033214.2 | NP_001028386.1 | 2 | 264 | ||
| GenBank mRNA | AK087874.1 | - | 2 | 76 | ||
| AK136036.1 | - | 2 | 265 | |||
| AK163316.1 | - | 2 | 56 | |||
| BC139082.1 | - | 2 | 235 | 8862104 | ||
| BC139083.1 | - | 2 | 235 | 8862105 | ||

E330034G19Rik
Genomic Map