siRNA Details
| Gene Symbol: Fam170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164485.1 | NP_001157957.1 | 2 | 1493 | ||
| GenBank mRNA | AK076630.1 | - | 1 | 2268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam170b| Gene Symbol: Fam170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164485.1 | NP_001157957.1 | 2 | 1493 | ||
| GenBank mRNA | AK076630.1 | - | 1 | 2268 | ||