siRNA Details
| Gene Symbol: A630023A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251843.1 | NP_001238772.1 | 1 | 50 | ||
| GenBank mRNA | AW989347.1 | - | 1 | 50 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630023A22Rik| Gene Symbol: A630023A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251843.1 | NP_001238772.1 | 1 | 50 | ||
| GenBank mRNA | AW989347.1 | - | 1 | 50 | ||