siRNA Details
| Gene Symbol: Zfp957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033215.3 | NP_001028387.1 | 1 | 112 | ||
| GenBank mRNA | AK135685.1 | - | 1 | 54 | ||
| AK136207.1 | - | 1 | 114 | |||
| AK139686.1 | - | 1 | 42 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp957| Gene Symbol: Zfp957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033215.3 | NP_001028387.1 | 1 | 112 | ||
| GenBank mRNA | AK135685.1 | - | 1 | 54 | ||
| AK136207.1 | - | 1 | 114 | |||
| AK139686.1 | - | 1 | 42 | |||