siRNA Details
| Gene Symbol: Zfp957 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033215.3 | NP_001028387.1 | 2 | 580 | ||
| GenBank mRNA | AK135685.1 | - | 2 | 522 | ||
| AK136207.1 | - | 2 | 583 | |||
| AK139686.1 | - | 2 | 510 | |||
| BC151187.1 | - | 1 | 174 | 9088076 | ||

Zfp957
Genomic Map