siRNA Details
| Gene Symbol: Dennd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081066.1 | NP_001074535.1 | 23 | 4160 | ||
| GenBank mRNA | AK173046.1 | - | 23 | 4060 | ||
| BC030441.1 | - | 3 | 579 | |||
| BC132389.1 | - | 23 | 4048 | 40130666 | ||
| BC137791.1 | - | 23 | 4048 | 8860810 | ||
| BC144912.1 | - | 24 | 4176 | |||

Dennd3
Genomic Map