siRNA Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039521.1 | NP_001034610.1 | 14 | 1365 | ||
| GenBank mRNA | AK080397.1 | - | 14 | 1321 | ||
| AK080680.1 | - | 1346 | ||||
| AK165676.1 | - | 14 | 1353 | |||
| BC034110.1 | - | 10 | 880 | |||
| BC138774.1 | - | 14 | 1330 | 8861796 | ||
| BC138776.1 | - | 14 | 1330 | 8861798 | ||

Rrn3
Genomic Map