siRNA Details
| Gene Symbol: Wdr90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163766.1 | NP_001157238.1 | 36 | 5042 | ||
| GenBank mRNA | AK014328.1 | - | 32 | 4143 | ||
| AK129465.1 | - | 35 | 4957 | |||
| AK154827.1 | - | 34 | 5208 | |||
| BC009168.1 | - | 5 | 694 | |||
| BC025468.1 | - | 10 | 1300 | |||
| BC043315.1 | - | 13 | 1733 | |||

Wdr90
Genomic Map